Learn More
Trajan Scientific SYRINGE REMOVE 50MM SK10R 6/PK
SDP
Catalog No. 502530405
Encompass_Preferred
SGE Series II Standard Removable Needle Syringes, Volume: 10 uL, Needle Gauge: 26 G, Diameter Outer Needle: 0.47 mm, 0.01 in., Quantity: 6/Pk., Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Removable, Diameter Outer: 0.25 in., 6.5 mm, Includes: Plunger protection
Catalog No. 50-253-0405 Supplier Trajan Scientific Supplier No. 002080
SGE Series II Standard Removable Needle Syringes, Volume: 10 uL, Needle Gauge: 26 G, Diameter Outer Needle: 0.47 mm, 0.01 in., Quantity: 6/Pk., Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Removable, Diameter Outer: 0.25 in., 6.5 mm, Includes: Plunger protection