Learn More
Trajan Scientific SYRINGE SHIMADZU 10R-S-0.47
SDP
Catalog No. 502530372
Encompass_Preferred
Trajan Autosampler Syringes, Connection Type: Removable, Volume: 10 uL, For Use With: Shimadzu Models AOC14, AOC17, AOC20, AOC20i, Gas chromatography, Needle Gauge: 26 G, Length Needle: 42 mm, 1.6 in., Diameter Outer Needle: 0.01 in., 0.47 mm, Length: 1.65 in., 42 mm, Model: 10R-S-0.47C
Catalog No. 50-253-0372 Supplier Trajan Scientific Supplier No. 002897
Trajan Autosampler Syringes, Connection Type: Removable, Volume: 10 uL, For Use With: Shimadzu Models AOC14, AOC17, AOC20, AOC20i, Gas chromatography, Needle Gauge: 26 G, Length Needle: 42 mm, 1.6 in., Diameter Outer Needle: 0.01 in., 0.47 mm, Length: 1.65 in., 42 mm, Model: 10R-S-0.47C