Learn More
Trajan Scientific SYRINGE SK 101N WG 10A 6/PK
SDP
Catalog No. 502550778
Encompass_Preferred
SGE Gastight Syringes: Fixed Needle Models, Volume: 10 uL, Needle Gauge: 26 G, Model: 10F-GT, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Fixed, Diameter Outer: 0.25 in., 6.5 mm, Diameter Outer Needle: 0.01 in., 0.47 mm
Catalog No. 50-255-0778 Supplier Trajan Scientific Supplier No. 002202
SGE Gastight Syringes: Fixed Needle Models, Volume: 10 uL, Needle Gauge: 26 G, Model: 10F-GT, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Fixed, Diameter Outer: 0.25 in., 6.5 mm, Diameter Outer Needle: 0.01 in., 0.47 mm