Learn More
Trajan Scientific SYRINGE SK-10F-HP-0.63 6/PK
SDP
Catalog No. 502550777
Encompass_Preferred
SGE Autosampler Syringes with 42mm Cone Tip Fixed Needle, Description: Not gas tight, Volume: 10 uL, Diameter Outer Needle: 0.63 mm, Needle Gauge: 23 G, Length Needle: 42 mm, Connection Type: Fixed, For Use With: Gas chromatography, Agilent GC Autosamplers HP7673, 7683, 7693A & 6850ALS
Catalog No. 50-255-0777 Supplier Trajan Scientific Supplier No. 002814
SGE Autosampler Syringes with 42mm Cone Tip Fixed Needle, Description: Not gas tight, Volume: 10 uL, Diameter Outer Needle: 0.63 mm, Needle Gauge: 23 G, Length Needle: 42 mm, Connection Type: Fixed, For Use With: Gas chromatography, Agilent GC Autosamplers HP7673, 7683, 7693A & 6850ALS