Learn More
Pluriselect Syringe Strainer 40 µm, Sterile, 12 per Pack
Catalog No. NC2748871
Encompass_Preferred
Sieving or filtration device for large quantities to exclude or concentrate particles from liquids. The strainer can be connected to a syringe or a tube via the screw cap with a Luer-Lock adaptor which works under positive or negative pressure.
Catalog No. NC2748871 Supplier Pluriselect Supplier No. 437104051
Sieving or filtration device for large quantities to exclude or concentrate particles from liquids. The strainer can be connected to a syringe or a tube via the screw cap with a Luer-Lock adaptor which works under positive or negative pressure.