Learn More
Med Vet International Exel Tuberculin Syringe, 1mL, Luer Lock, 100/Box, 26049
Catalog No. 502522568
Encompass_Preferred
Exel International 1mL Syringe comes with a clear barrel and easy to read graduation marks. It has a low dead space plunger. The Luer Lock tip of this 1mL syringe fits conventional and safety needles. This quantity is 100/BX
Catalog No. 50-252-2568 Supplier Med Vet International Supplier No. 26049
 Restricted Item
Add to Cart
Add to Cart
Exel International 1mL Syringe comes with a clear barrel and easy to read graduation marks. It has a low dead space plunger. The Luer Lock tip of this 1mL syringe fits conventional and safety needles. This quantity is 100/BX