Learn More
Biotang Inc E-Z Syringe tips 1.25ML Non-STERILE, 100/PK
Supplier:  Biotang Inc 702373
Encompass_Preferred
Fit most manufacture's repeating pipettes using typical standard sized syringe tips, doesn't fit Eppendorf models. Made in Germany.
Catalog No. 50-751-5011