Learn More
Biotang Inc E-Z Syringe Tip 5.0ML, Sterile, 100pk
Supplier:  Biotang Inc 702391
Encompass_Preferred
Fit most manufacture's repeating pipettes using typical standard sized syringe tips, doesn't fit Eppendorf models. Made in Germany.
Catalog No. 50-751-5006