Learn More
Biotang Inc E-Z Syringe tips 50ML STERILE
Supplier:  Biotang Inc 702397
Encompass_Preferred
Fit most manufacture's repeating pipettes using typical standard sized syringe tips, doesn't fit Eppendorf models. Made in Germany.
Catalog No. 50-751-5009