Learn More
Electron Microscopy Sciences TorrLube Syringe 1 CM3
SDP
Catalog No. 5034118
Encompass_Preferred
TorrLube Extreme Grease is white, odorless, nonflammable, and chemically inert products based on TorrLube Oil and the most advanced PTFE thickener available. It does not contain Sodium or Iron. TorrLube Extreme Grease remains stable and will not break down under mechanical stress and lose its ability to hold TorrLube Oil. TorrLube Extreme Grease helps prevent sticking, jumping, and mechanical noise, securing smooth, controlled movement. Syringes have a #22 gauge blunt needle.
Catalog No. 50-341-18 Supplier Electron Microscopy Sciences Supplier No. 6075001
TorrLube Extreme Grease is white, odorless, nonflammable, and chemically inert products based on TorrLube Oil and the most advanced PTFE thickener available. It does not contain Sodium or Iron. TorrLube Extreme Grease remains stable and will not break down under mechanical stress and lose its ability to hold TorrLube Oil. TorrLube Extreme Grease helps prevent sticking, jumping, and mechanical noise, securing smooth, controlled movement. Syringes have a #22 gauge blunt needle.