Learn More
Waters Corp Waters Syringe Valve, 4-Port, for 2707 Autosampler
Catalog No. 50792918 Shop All Waters Corporation Products
Encompass_Preferred
This replacement syringe valve is designed for the 2707 autosampler. It functions as a 4-port selection valve, directing liquids for various functions including waste, wash, and connecting to the buffer tubing.
  • Replacement valve for the 2707 autosampler.
  • Features a 4-port selection design, with one port unused.
  • Includes a dedicated “waste” port for draining the syringe dispenser.
  • Equipped with a “wash” port for aspirating wash liquid from a wash bottle.
  • Connects to buffer tubing via the “needle” port.
  • Suitable for HPLC systems.
Catalog No. 50-792-918 Supplier Waters Corp Supplier No. 700003846
This replacement syringe valve is designed for the 2707 autosampler. It functions as a 4-port selection valve, directing liquids for various functions including waste, wash, and connecting to the buffer tubing.
  • Replacement valve for the 2707 autosampler.
  • Features a 4-port selection design, with one port unused.
  • Includes a dedicated “waste” port for draining the syringe dispenser.
  • Equipped with a “wash” port for aspirating wash liquid from a wash bottle.
  • Connects to buffer tubing via the “needle” port.
  • Suitable for HPLC systems.