Learn More
Trajan Scientific Syringe Valve Medical Lab 1/EA
SDP
Catalog No. 5000493836
Encompass_Preferred
Push-pull valve with removable 5 cm 0 63 mm OD cone tip needle for 25 L to 2 5 mL removable needle syringes
Catalog No. 50-004-93836 Supplier Trajan Scientific Supplier No. 031905
Push-pull valve with removable 5 cm 0 63 mm OD cone tip needle for 25 L to 2 5 mL removable needle syringes