Learn More
Trajan Scientific SYRINGE VALVE VVLMA
SDP
Catalog No. 502530274
Encompass_Preferred
Trajan Replacement Valves, For Use With: 5mL to 2L syringe volume, Description: Push buttone with Luer Lock
Catalog No. 50-253-0274 Supplier Trajan Scientific Supplier No. 031910
Trajan Replacement Valves, For Use With: 5mL to 2L syringe volume, Description: Push buttone with Luer Lock