Learn More
Electron Microscopy Sciences Syringe With Tips 0.6 ml, Autoclavable
SDP
Catalog No. 501899951
Encompass_Preferred
Used with our repetitive syringe dispenser for dispensing multiple doses of a set volume. The 0.6 ml syringe dispenses 10 to 20 uL.
Type: Autoclavable
Catalog No. 50-189-9951 Supplier Electron Microscopy Sciences Supplier No. 72482
Used with our repetitive syringe dispenser for dispensing multiple doses of a set volume. The 0.6 ml syringe dispenses 10 to 20 uL.
Type: Autoclavable