Learn More
Electron Microscopy Sciences Syringe With Tips, 1.5 ml, Autoclavable
SDP
Catalog No. 501899952
Encompass_Preferred
Used with our repetitive syringe dispenser for dispensing multiple doses of a set volume. The 1.5 ml syringe dispenses 25 to 125 uL.
Type: Autoclavable
Catalog No. 50-189-9952 Supplier Electron Microscopy Sciences Supplier No. 72483
Used with our repetitive syringe dispenser for dispensing multiple doses of a set volume. The 1.5 ml syringe dispenses 25 to 125 uL.
Type: Autoclavable