Learn More
Electron Microscopy Sciences Rubber Piston Syringes with 25 gauge Needle 10 ml
SDP
Catalog No. 50949327
Encompass_Preferred
These high quality disposable rubber piston syringes offer substantial savings. A lubricated synthetic rubber piston allows for a smooth draw while a positive stop prevents accidental pull out. Bold graduations are easily read and resist wipe-off. Syringes are ETO gas sterilized, latex and pyrogen free. All syringes are packaged in individual sterile strip packs. Needle is 5/8".
Volume: 1 ml
Catalog No. 50-949-327 Supplier Electron Microscopy Sciences Supplier No. 72515
These high quality disposable rubber piston syringes offer substantial savings. A lubricated synthetic rubber piston allows for a smooth draw while a positive stop prevents accidental pull out. Bold graduations are easily read and resist wipe-off. Syringes are ETO gas sterilized, latex and pyrogen free. All syringes are packaged in individual sterile strip packs. Needle is 5/8".
Volume: 1 ml