Learn More
Med Vet International Exel Syringe & Needle, 10mL, Luer Lock, 21G X 1in, Hypodermic, 100/BX, 26252
Supplier:  Med Vet International 26252
Encompass_Preferred
Exel International 10mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 10mL syringe fits conventional and safety needles. This Exel International 26252 Syringe & Needle Combo comes with a 21G X 1' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-255-9752