Learn More
Med Vet International AHS Tuberculin Syringe & Needle, 1mL, Luer Slip, 27 X 1/2in, 100/BX, AH01T2713
Catalog No. 502559607
Encompass_Preferred
AHS AH01T2713 1mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 1mL syringe fits conventional and safety needles. This AHS Needle is a 27 X 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-255-9607 Supplier Med Vet International Supplier No. AH01T2713
AHS AH01T2713 1mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 1mL syringe fits conventional and safety needles. This AHS Needle is a 27 X 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX