Learn More
Med Vet International Terumo Syringe, 30mL, Luer Lock, 25/Box, SS-30L
Catalog No. 502092991
Encompass_Preferred
Terumo SS-30L 30mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 30mL syringe fits conventional and safety needles. This quantity is 25/BX
Catalog No. 50-209-2991 Supplier Med Vet International Supplier No. 30L
Terumo SS-30L 30mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 30mL syringe fits conventional and safety needles. This quantity is 25/BX