Learn More
Med Vet International Exel Syringe & Needle, 3mL, Luer Lock, 20G X 1 1/2in, Hypodermic, 100/BX, 26109
Catalog No. 502092999
Encompass_Preferred
Exel International 3mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This Exel International 26109 Syringe & Needle Combo comes with a 20G X 1 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
This item is not returnable. View return policy
Catalog No. 50-209-2999 Supplier Med Vet International Supplier No. 26109
Exel International 3mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This Exel International 26109 Syringe & Needle Combo comes with a 20G X 1 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX