Learn More
Med Vet International Exel Syringe & Needle, 3mL, Luer Slip, 20G x in, Hypodermic, 100/Box
Catalog No. 502093000
Encompass_Preferred
Exel International 3mL syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This Exel International 26611 Syringe & Needle Combo comes with a 20G x 1" regular wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.
This item is not returnable. View return policy
Catalog No. 50-209-3000 Supplier Med Vet International Supplier No. 26611
Exel International 3mL syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This Exel International 26611 Syringe & Needle Combo comes with a 20G x 1" regular wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.