Learn More
Med Vet International Exel Syringe & Needle, 3mL, Luer Lock, 21G X 1 1/4in, Hypodermic, 100/BX, 26127
Supplier:  Med Vet International 26127
Encompass_Preferred
Exel International 3mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This Exel International 26127 Syringe & Needle Combo comes with a 21G X 1 1/4' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-255-9757
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.