Learn More
Med Vet International Exel International Luer Slip Syringe & Needle Combo, 3mL, 21G x 1in Regular Wall, Hypodermic, 100/Box 26608
Supplier:  Med Vet International 26608
Encompass_Preferred
Exel International 3mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This Exel International 26608 Syringe & Needle Combo comes with a 21G X 1' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-255-9730