Learn More
Med Vet International Exel Syringe & Needle, 3mL, Luer Lock, 21G X 1.5in, Hypodermic, 100/BX, 26107
Catalog No. 502093009
Encompass_Preferred
Exel International 3mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This Exel International 26107 Syringe & Needle Combo comes with a 21G X 1.5' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
This item is not returnable. View return policy
Catalog No. 50-209-3009 Supplier Med Vet International Supplier No. 26107
Exel International 3mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This Exel International 26107 Syringe & Needle Combo comes with a 21G X 1.5' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX