Learn More
Med Vet International Terumo Syringe & Needle, 3mL, Luer Lock, 22GX 1in, Hypodermic, 100/BX, SS-03L2225
Catalog No. 502093010
Encompass_Preferred
Terumo SS-03L2225 3mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This Terumo SS-03L2225 3mL Syringe & Needle Combo comes with a 22GX 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This quantity is 100/BX
Catalog No. 50-209-3010 Supplier Med Vet International Supplier No. 03L2225
Terumo SS-03L2225 3mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This Terumo SS-03L2225 3mL Syringe & Needle Combo comes with a 22GX 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This quantity is 100/BX