Learn More
Med Vet International Exel Syringe & Needle, 3mL, Luer Slip, 22G X 3/4in, Hypodermic, 100/BX, 26601
Catalog No. 502559733
Encompass_Preferred
Exel International 3mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 3mL syringe fits conventional and safety needles. This Exel International 26601 Syringe & Needle Combo comes with a 22G X 3/4' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-255-9733 Supplier Med Vet International Supplier No. 26601
Exel International 3mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 3mL syringe fits conventional and safety needles. This Exel International 26601 Syringe & Needle Combo comes with a 22G X 3/4' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX