Learn More
Med Vet International Nipro Syringe & Needle, 3mL, Luer Slip, 22g X 1", Hypodermic, 100/Box, Jd+03S2225
Catalog No. 502093015
Encompass_Preferred
Nipro JD+03S2225 3mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 3mL syringe fits conventional and safety needles. This Nipro JD+03S2225 Syringe & Needle Combo comes with a 22G X 1" Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles allow rapid and efficient drug delivery. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-209-3015 Supplier Med Vet International Supplier No. JD03S2225
Nipro JD+03S2225 3mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 3mL syringe fits conventional and safety needles. This Nipro JD+03S2225 Syringe & Needle Combo comes with a 22G X 1" Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles allow rapid and efficient drug delivery. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX