Learn More
Med Vet International Terumo Syringe & Needle, 3mL, Luer Lock, 23GX 1 1/2in, Hypodermic, 100/BX, SS-03L2338
Supplier:  Med Vet International 03L2338
Encompass_Preferred
Terumo SS-03L2338 3mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This Terumo SS-03L2338 3mL Syringe & Needle Combo comes with a 23GX 1 1/2' Thin Wall needle. Terumo's thin wall needles areprecisely honed and beveled. This makes for ease of penetration and less patient discomfort This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-255-9763