Learn More
Med Vet International Exel Syringe, 50-60mL, Luer Lock, 25/Box, 26300
Supplier:  Med Vet International 26300
Encompass_Preferred
Exel International 50-60mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 50-60mL syringe fits conventional and safety needles. This quantity is 25/BX
Catalog No. 50-209-3046