Learn More
Med Vet International Nipro Syringe, 50mL, Luer Slip, 50/Box, Jd+50Es-G
Catalog No. 502093047
Encompass_Preferred
Nipro JD+50ES-G 50mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 50mL syringe fits conventional and safety needles. This quantity is 25/BX
Catalog No. 50-209-3047 Supplier Med Vet International Supplier No. JD50ES
Nipro JD+50ES-G 50mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 50mL syringe fits conventional and safety needles. This quantity is 25/BX