Learn More
Med Vet International Exel Syringe & Needle, 5mL, Luer Lock, 21G X 1 1/2in, Hypodermic, 100/BX, 26213
Catalog No. 502093053
Encompass_Preferred
Exel International 5mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 5mL syringe fits conventional and safety needles. This Exel International 26213 Syringe & Needle Combo comes with a 21G X 1 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
This item is not returnable. View return policy
Catalog No. 50-209-3053 Supplier Med Vet International Supplier No. 26213
Exel International 5mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 5mL syringe fits conventional and safety needles. This Exel International 26213 Syringe & Needle Combo comes with a 21G X 1 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX