Learn More
Med Vet International Nipro Syringe, 5mL, Luer Slip, 100/Box, Jd+05S
Catalog No. 502093062
Encompass_Preferred
Nipro JD+05S 5mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 5mL syringe fits conventional and safety needles. This quantity is 100/BX
Catalog No. 50-209-3062 Supplier Med Vet International Supplier No. JD05S
Nipro JD+05S 5mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 5mL syringe fits conventional and safety needles. This quantity is 100/BX