Learn More
Med Vet International Terumo Syringe, 5mL, Luer Slip, 100/Box, SS-05S
Catalog No. 502093064
Encompass_Preferred
Terumo SS-05S 5mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 5mL syringe fits conventional and safety needles. This quantity is 100/BX
Catalog No. 50-209-3064 Supplier Med Vet International Supplier No. 05S
Terumo SS-05S 5mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 5mL syringe fits conventional and safety needles. This quantity is 100/BX