Learn More
Med Vet International AHS Syringe, 60mL, Luer Slip, 25/Box, AH60S
Supplier:  Med Vet International AH60S
Encompass_Preferred
AHS AH60S 60mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 60mL syringe fits conventional and safety needles. This quantity is 25/BX
Catalog No. 50-209-3066