Learn More
Med Vet International Exel Syringe & Needle, 1mL, Permanent Needle, 28G X 1/2in, Insulin, 100/BX, 26027
Catalog No. 502093070
Encompass_Preferred
In convenient packages of 10. Exel International 1mL Syringe comes with a clear barrel and easy to read graduation marks. This syringe contains a permanently attached needle. This Exel International 26027 Syringe & Needle Combo comes with a 28G X 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is an insulin syringe. It is used to inject insulin in the subcutaneous fat. This quantity is 100/BX
This item is not returnable. View return policy
Catalog No. 50-209-3070 Supplier Med Vet International Supplier No. 26027
In convenient packages of 10. Exel International 1mL Syringe comes with a clear barrel and easy to read graduation marks. This syringe contains a permanently attached needle. This Exel International 26027 Syringe & Needle Combo comes with a 28G X 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is an insulin syringe. It is used to inject insulin in the subcutaneous fat. This quantity is 100/BX