Learn More
Med Vet International AHS Syringes, 20mL Luer Lock, 50/Box, AH20L
Catalog No. 502522466
Encompass_Preferred
AHS AH20L 20mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer lock tip of this 20mL syringe fits conventional and safety needles. This quantity is 50/BX
Catalog No. 50-252-2466 Supplier Med Vet International Supplier No. AH20L
AHS AH20L 20mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer lock tip of this 20mL syringe fits conventional and safety needles. This quantity is 50/BX