Learn More
Med Vet International AHS Syringes, 20mL Luer Slip, 50/Box, AH20S
Catalog No. 502522465
Encompass_Preferred
AHS AH20S 20mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 20mL syringe fits conventional and safety needles. This quantity is 50/BX
Catalog No. 50-252-2465 Supplier Med Vet International Supplier No. AH20S
AHS AH20S 20mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 20mL syringe fits conventional and safety needles. This quantity is 50/BX