Learn More
Instech Sterile Catheter Flush/Lock Sl
SDP
Catalog No. 5000510047
Encompass
The Instech USP-HS-020-0 5-NJ1-50 is a sterile catheter flush/lock solution formulated with heparin at 20 USP units per mL in 0 9% saline It is supplied pre-filled to 0 5 mL in an HSW NORM-JECT 1 mL syringe with a slip-fit cap sans PNP3M PinPort injector The syringes are sealed to maintain sterility and provided in 10 sterile pouches of 5 (50 syringes) br br Features ul li Heparin Concentration 20 USP units/mL /li li Saline Concentration 0 9% NaCl /li li Fill Volume 0 5 mL /li li Syringe Type HSW NORM-JECT 1 mL slip fit syringe /li li Closure Slip-fit cap (no PNP3M PinPort injector) /li li Sterility USP-standard sterile compounding /li li Package Configuration 10 pouches of 5 (50 syringes) /li li Use Case Catheter flush/lock solution for maintaining catheter patency in laboratory animals not for human use /li /ul
Catalog No. 50-005-10047 Supplier Instech Supplier No. USPHS0200.5NJ150
The Instech USP-HS-020-0 5-NJ1-50 is a sterile catheter flush/lock solution formulated with heparin at 20 USP units per mL in 0 9% saline It is supplied pre-filled to 0 5 mL in an HSW NORM-JECT 1 mL syringe with a slip-fit cap sans PNP3M PinPort injector The syringes are sealed to maintain sterility and provided in 10 sterile pouches of 5 (50 syringes) br br Features ul li Heparin Concentration 20 USP units/mL /li li Saline Concentration 0 9% NaCl /li li Fill Volume 0 5 mL /li li Syringe Type HSW NORM-JECT 1 mL slip fit syringe /li li Closure Slip-fit cap (no PNP3M PinPort injector) /li li Sterility USP-standard sterile compounding /li li Package Configuration 10 pouches of 5 (50 syringes) /li li Use Case Catheter flush/lock solution for maintaining catheter patency in laboratory animals not for human use /li /ul