Learn More
Electron Microscopy Sciences Rubber Piston Syringes without Needles 30 ml, Centric
SDP
Catalog No. 50949463
Encompass_Preferred
These high quality disposable rubber piston syringes offer substantial savings. A lubricated synthetic rubber piston allows for a smooth draw while a positive stop prevents accidental pull out. Bold graduations are easily read and resist wipe-off. Syringes are ETO gas sterilized, latex and pyrogen free. All syringes are packaged in individual sterile strip packs.
Volume: 30 ml
Type: Centric
Catalog No. 50-949-463 Supplier Electron Microscopy Sciences Supplier No. 72513LL
These high quality disposable rubber piston syringes offer substantial savings. A lubricated synthetic rubber piston allows for a smooth draw while a positive stop prevents accidental pull out. Bold graduations are easily read and resist wipe-off. Syringes are ETO gas sterilized, latex and pyrogen free. All syringes are packaged in individual sterile strip packs.
Volume: 30 ml
Type: Centric