Learn More
Electron Microscopy Sciences Rubber Piston Syringes with 21 gauge Needle 10 ml
SDP
Catalog No. 50949404
Encompass_Preferred
These high quality disposable rubber piston syringes offer substantial savings. A lubricated synthetic rubber piston allows for a smooth draw while a positive stop prevents accidental pull out. Bold graduations are easily read and resist wipe-off. Syringes are ETO gas sterilized, latex and pyrogen free. All syringes are packaged in individual sterile strip packs. Neeedle is 1 1/2".
Volume: 10 ml
Catalog No. 50-949-404 Supplier Electron Microscopy Sciences Supplier No. 72518
These high quality disposable rubber piston syringes offer substantial savings. A lubricated synthetic rubber piston allows for a smooth draw while a positive stop prevents accidental pull out. Bold graduations are easily read and resist wipe-off. Syringes are ETO gas sterilized, latex and pyrogen free. All syringes are packaged in individual sterile strip packs. Neeedle is 1 1/2".
Volume: 10 ml