Learn More
Med Vet International AHS Syringes, 60mL Luer Lock, 25/Box, AH60L
Catalog No. 502093079
Encompass_Preferred
AHS AH60L 60mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer lock tip of this 60mL syringe fits conventional and safety needles. This quantity is 25/BX
Catalog No. 50-209-3079 Supplier Med Vet International Supplier No. AH60L
AHS AH60L 60mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer lock tip of this 60mL syringe fits conventional and safety needles. This quantity is 25/BX