Learn More
Medchemexpress LLC Syringetin-3-O-glucoside | 40039-49-4 | 1 MG
SDP
Catalog No. 5000417387
Encompass_Preferred
Syringetin-3-O-glucoside (Syringetin 3-O-beta-D-glucoside) is a flavonol glycoside that exhibits relatively weak DPPH and ABTS radical scavenging activity.
  • Exhibits antioxidant activities.
Catalog No. 50-004-17387 Supplier Medchemexpress LLC Supplier No. HYN81941MG
Syringetin-3-O-glucoside (Syringetin 3-O-beta-D-glucoside) is a flavonol glycoside that exhibits relatively weak DPPH and ABTS radical scavenging activity.
  • Exhibits antioxidant activities.