Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Syringetin-3-O-glucoside | 40039-49-4 | 10 MG
SDP

Catalog No. 5000419145
Encompass_Preferred

Syringetin-3-O-glucoside (Syringetin 3-O-β-D-glucoside) is a flavonol glycoside that shows relatively weak DPPH and ABTS radical scavenging activity. It is intended for research use only.

  • Purity: 99.37%
  • Molecular weight: 508.43
  • Chemical formula: C23H24O13
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Classification: Flavonoids, Flavonols, Phenols, Polyphenols
  • Initial source: Plants, Vitaceae, Vitis vinifera cv. Zalema
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month (protect from light)
  • Solubility (in vitro): Ethanol < 1 mg/mL (insoluble)

Catalog No. 50-004-19145 Supplier Medchemexpress LLC Supplier No. HYN819410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Syringetin-3-O-glucoside (Syringetin 3-O-β-D-glucoside) is a flavonol glycoside that shows relatively weak DPPH and ABTS radical scavenging activity. It is intended for research use only.

  • Purity: 99.37%
  • Molecular weight: 508.43
  • Chemical formula: C23H24O13
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Classification: Flavonoids, Flavonols, Phenols, Polyphenols
  • Initial source: Plants, Vitaceae, Vitis vinifera cv. Zalema
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month (protect from light)
  • Solubility (in vitro): Ethanol < 1 mg/mL (insoluble)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.