Learn More
Medchemexpress LLC HY-N0339 100mg Medchemexpress, Syringic acid CAS:530-57-4 Purity:>98%
SDP
Catalog No. 501933527
Encompass_Preferred
Medchemexpress, HY-N0339 100mg Syringic acid CAS:530-57-4 Syringic acid is correlated with high antioxidant activity and inhibition of LDL oxidation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3527 Supplier Medchemexpress LLC Supplier No. HYN0339100MG
Medchemexpress, HY-N0339 100mg Syringic acid CAS:530-57-4 Syringic acid is correlated with high antioxidant activity and inhibition of LDL oxidation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.