Learn More
Indofine Chemical Syringic Acid, 99%, 25 Gm
SDP
Catalog No. 50908306
Encompass_Preferred
SYRINGIC ACID 25 GM, Rare Organics & Biochemicals, 4-Hydroxy-3,5-dimethoxybenzoic acid, C9H10O5, MW: 198.18, 99+%, 530-57-4, MFCD00002552
Catalog No. 50-908-306 Supplier Indofine Chemical Supplier No. 026107 25 GM
SYRINGIC ACID 25 GM, Rare Organics & Biochemicals, 4-Hydroxy-3,5-dimethoxybenzoic acid, C9H10O5, MW: 198.18, 99+%, 530-57-4, MFCD00002552