Learn More
Medchemexpress LLC Syringic acid | 530-57-4 | 99.58% | 198.17 | 500 MG
SDP
Catalog No. 5000427213
Encompass_Preferred
Syringic acid is correlated with high antioxidant activity and inhibition of LDL oxidation. It is a phenol present in some distilled alcohol beverages and is also a product of microbial (gut) metabolism of anthocyanins and other polyphenols that have been consumed. Research suggests that phenolics from wine, such as Syringic acid, may play a positive role against the oxidation of low-density lipoprotein (LDL), which is a key step in the development of atherosclerosis.
  • High antioxidant activity
  • Inhibits LDL oxidation
  • Found in some distilled alcohol beverages
  • Product of microbial metabolism of polyphenols
  • May help prevent atherosclerosis
  • Solid appearance
  • White to off-white color
  • Soluble in DMSO
Catalog No. 50-004-27213 Supplier Medchemexpress LLC Supplier No. HYN0339500MG
Add to Cart
Add to Cart
Syringic acid is correlated with high antioxidant activity and inhibition of LDL oxidation. It is a phenol present in some distilled alcohol beverages and is also a product of microbial (gut) metabolism of anthocyanins and other polyphenols that have been consumed. Research suggests that phenolics from wine, such as Syringic acid, may play a positive role against the oxidation of low-density lipoprotein (LDL), which is a key step in the development of atherosclerosis.
  • High antioxidant activity
  • Inhibits LDL oxidation
  • Found in some distilled alcohol beverages
  • Product of microbial metabolism of polyphenols
  • May help prevent atherosclerosis
  • Solid appearance
  • White to off-white color
  • Soluble in DMSO