Learn More
TARGETMOL CHEMICALS INC SYRINGIN 25MG
SDP
Catalog No. 502576949
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg and bulk. Please contact Fisher for quotes. Syringin (eleutheroside-b) is a compound with antitumor antiproliferative immunomodulatory and platelet aggregation inhibiting effects. purity: 99%
Catalog No. 50-257-6949 Supplier TARGETMOL CHEMICALS INC Supplier No. T340825MG
Also available in 5 mg 10 mg 50 mg and bulk. Please contact Fisher for quotes. Syringin (eleutheroside-b) is a compound with antitumor antiproliferative immunomodulatory and platelet aggregation inhibiting effects. purity: 99%