Learn More
Medchemexpress LLC HY-N0824 5mg Medchemexpress, Syringin CAS:118-34-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN08245MG
Encompass_Preferred
Medchemexpress, HY-N0824 5mg Syringin CAS:118-34-3 Syringin is a main bioactive phenolic glycoside in Acanthopanax senticosus, with anti-osteoporosis activity. Syringin prevents cardiac hypertrophy induced by pressure overload through the attenuation of autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2824