Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Syringin (eleutheroside B) | 118-34-3 | MFCD00016778 | >93.0% | 372.37 | C17H24O9 | 10 MG
SDP

Catalog No. 5000296426
Encompass_Preferred

Syringin (eleutheroside B) is a natural phenolic glycoside used in research to probe autophagy-related pathways and other bioactivities such as anti-osteoporosis and cardioprotection. Supplied as an analytical research quantity, the compound has formula C17H24O9 and molecular weight 372.37 g/mol.

  • Suitable for in vitro and analytical applications.
  • Soluble in DMSO; high-concentration stock solutions are achievable.
  • Documented storage: store at 4°C and protect from light; in solvent, -80°C for long-term storage.
  • Molecular formula and weight provided for formulation and dosing calculations.

Catalog No. 50-002-96426 Supplier Medchemexpress LLC Supplier No. HYN082410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Syringin (eleutheroside B) is a natural phenolic glycoside used in research to probe autophagy-related pathways and other bioactivities such as anti-osteoporosis and cardioprotection. Supplied as an analytical research quantity, the compound has formula C17H24O9 and molecular weight 372.37 g/mol.

  • Suitable for in vitro and analytical applications.
  • Soluble in DMSO; high-concentration stock solutions are achievable.
  • Documented storage: store at 4°C and protect from light; in solvent, -80°C for long-term storage.
  • Molecular formula and weight provided for formulation and dosing calculations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.