Learn More
Medchemexpress LLC Syringin (standard) | 118-34-3 | 372.37 | 20 MG
SDP
Catalog No. 5000415893
Encompass_Preferred
Syringin is a primary bioactive phenolic glycoside found in *Acanthopanax senticosus*. It exhibits anti-osteoporosis activity and may prevent cardiac hypertrophy induced by pressure overload by attenuating autophagy.
  • Main bioactive phenolic glycoside
  • Exhibits anti-osteoporosis activity
  • Prevents cardiac hypertrophy by attenuating autophagy
  • Decreases hypertrophic markers and increases α-MHC levels in cells
  • Improves bone mineral density and bone microarchitecture in vivo
Catalog No. 50-004-15893 Supplier Medchemexpress LLC Supplier No. HYN082420MG
Syringin is a primary bioactive phenolic glycoside found in *Acanthopanax senticosus*. It exhibits anti-osteoporosis activity and may prevent cardiac hypertrophy induced by pressure overload by attenuating autophagy.
  • Main bioactive phenolic glycoside
  • Exhibits anti-osteoporosis activity
  • Prevents cardiac hypertrophy by attenuating autophagy
  • Decreases hypertrophic markers and increases α-MHC levels in cells
  • Improves bone mineral density and bone microarchitecture in vivo